Learn More
Cayman Chemical 10oxo12 Z 15 ZOctadecadI 100ug
SDP
Catalog No. 502919474
Encompass_Preferred
An oxylipin gut microbiota metabolite; decreases T0901317-induced increases in mRNA expression levels of the gene encoding SREBP-1c in HepG2 cells
Catalog No. 50-291-9474 Supplier Cayman Chemical Supplier No. 36976100 UG
An oxylipin gut microbiota metabolite; decreases T0901317-induced increases in mRNA expression levels of the gene encoding SREBP-1c in HepG2 cells