Learn More
Dojindo Molecular Technologies Inc Cellular Senescence Detection Kit (10 plate), detects SA-β-gal with high sensitivity by SPiDER-βGal, applicable to both living and fixed cells, by fluorescence microscopy or flow cytometry.
SDP
Catalog No. NC1961184
Encompass
SPiDER-βGal is a new reagent to detect β-galactosidase specifically which possesses a high cell-permeability and a high retentivity inside cells. This novel reagent can detect β-galactosidase not only in fixed cells but also in living cells, and also can be applied to quantitative analysis by flow cytometry. Senescence-associated beta-galactosidase (SA-β-gal), which is overexpressed in senescent cells, has been widely used as a marker of cellular senescence. Cellular Senescence Detection Kit – SPiDER-βGal is optimized to detect intracellular SA-β-gal. Modern publications with a high number of citations summarize the various features of cellular senescence on the following page (https://www.dojindo.com/senescent-cell-detection-selection-guide/).
Catalog No. NC1961184 Supplier Dojindo Molecular Technologies Inc Supplier No. SG0410
SPiDER-βGal is a new reagent to detect β-galactosidase specifically which possesses a high cell-permeability and a high retentivity inside cells. This novel reagent can detect β-galactosidase not only in fixed cells but also in living cells, and also can be applied to quantitative analysis by flow cytometry. Senescence-associated beta-galactosidase (SA-β-gal), which is overexpressed in senescent cells, has been widely used as a marker of cellular senescence. Cellular Senescence Detection Kit – SPiDER-βGal is optimized to detect intracellular SA-β-gal. Modern publications with a high number of citations summarize the various features of cellular senescence on the following page (https://www.dojindo.com/senescent-cell-detection-selection-guide/).