Learn More
Cayman Chemical 10PnxtrIfluoroacetAT salt 10mg
SDP
Catalog No. 502909320
Encompass_Preferred
A peptide inhibitor of PANX1; inhibits purinergic P2X7 receptor activation-induced dye uptake, constitutive dye uptake, and hemichannel currents, as well as ATP-induced caspase activation and IL-1β release in HEK293 cells at 200 µM; inhibits pannexin channel currents and connexin 46 channel currents in Xenopus oocytes expressing the human channels
Catalog No. 50-290-9320 Supplier Cayman Chemical Supplier No. 3686710 MG
A peptide inhibitor of PANX1; inhibits purinergic P2X7 receptor activation-induced dye uptake, constitutive dye uptake, and hemichannel currents, as well as ATP-induced caspase activation and IL-1β release in HEK293 cells at 200 µM; inhibits pannexin channel currents and connexin 46 channel currents in Xenopus oocytes expressing the human channels