Learn More
Perkin Elmer US LLC 10-Position Micro Sample Rack - 30 mm, 2x5
Catalog No. 502117216
Encompass_Preferred
This rack can be used with 15 mL Graduated PFA Vials (N8145132) and 30 mL Graduated PFA Vials (N8145133).
Catalog No. 50-211-7216 Supplier Perkin Elmer US LLC Supplier No. N8122463
This rack can be used with 15 mL Graduated PFA Vials (N8145132) and 30 mL Graduated PFA Vials (N8145133).