Learn More
SPEX CERTIPREP LLC 3P 10PPM HG IN 5%HNO VOLUME 10
Supplier:  SPEX CERTIPREP LLC 3P 10331100
Encompass_Preferred
(HPS by Zeptometrix) 10 ug/mL Mercury in 5% HNO3 Exp Date: 1 Year Exp Date: 12 months This standard is traceable to NIST and certified Guide 34 and 17025
Catalog No. NC9877553
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.