Learn More
SPECTRUM QUALITY STANDARDS UOP163 [10 ppm wt.] (6×20 ml)
Supplier:  SPECTRUM QUALITY STANDARDS UOP16310
Encompass_Preferred
Standards are prepared in a mixture of Isooctane and Toluene to determine inflection points during the Potentiometric Titration of Mercaptan Sulfur in Hydrocarbon liquids. The standards are prepared with Tert-Nonyl Mercaptan. They are packaged in 20 ml flame sealed ampoules and offered in various concentrations and other sizes.
Catalog No. NC1255630