Learn More
Aqua Solutions 10PPM TOC STANDARD 500ML
SDP
Catalog No. NC9566096
Encompass_Preferred
Total Organic Carbon Standard, 10.0 ppm, 500 mL, MP: ~0degreeC, Specific Gravity: 1,BP: ~100degreeC, Volatile by Volume: > 99%, Water Soluble, Appearance and Odors: Clear water white Odorless Liquid.
Catalog No. NC9566096 Supplier Aqua Solutions Supplier No. 9564500ML
Total Organic Carbon Standard, 10.0 ppm, 500 mL, MP: ~0degreeC, Specific Gravity: 1,BP: ~100degreeC, Volatile by Volume: > 99%, Water Soluble, Appearance and Odors: Clear water white Odorless Liquid.