Learn More
Trajan Scientific 10R-RSH-5.7/0.47C 10UL SYRINGE
SDP
Catalog No. 502530373
Encompass_Preferred
Trajan Autosampler Syringes With Removable Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.18 in., 0.47 mm, Needle Gauge: 26 G, Length Needle: 2.24 in., 57 mm, Model: 10R-RTC/RSH-5.7/0.47C, Volume: 10 uL, Connection Type: Removable
Catalog No. 50-253-0373 Supplier Trajan Scientific Supplier No. 002875
Trajan Autosampler Syringes With Removable Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.18 in., 0.47 mm, Needle Gauge: 26 G, Length Needle: 2.24 in., 57 mm, Model: 10R-RTC/RSH-5.7/0.47C, Volume: 10 uL, Connection Type: Removable