Learn More
Andwin Scientific 10UL AERO BAR TIP RKD ST 960/P
SDP
Catalog No. NC0812497
Encompass_Preferred
The 1000 L Barrier Pipet Tips by Neptune are specially designed with high density polyethlene filters in order to avoid contamination from aerosol. This product is sterile, is free of RNase, DNase, pyrogens, and PCR Inhibitors, and is made to fit with any standard size pipettors. It is sold in a pack of 3072 tips.
Catalog No. NC0812497 Supplier Andwin Scientific Supplier No. BT10XL
The 1000 L Barrier Pipet Tips by Neptune are specially designed with high density polyethlene filters in order to avoid contamination from aerosol. This product is sterile, is free of RNase, DNase, pyrogens, and PCR Inhibitors, and is made to fit with any standard size pipettors. It is sold in a pack of 3072 tips.