Learn More
Alkali Scientific Alkali Scientific™ 10uL Beckman™ Biomek FX/NX, Multimek and i Series Compatible Robotic Automation Tips, Filtered, 96-Well Head/Tip Format, Clear, Low Retention, Sterile, 5 Packs of 960, 4800/Case
SDP
Catalog No. 503026947
Encompass_Preferred
Crafted from high-quality polypropylene, Alkali Scientific™ provides a wide range of tips for Beckman Coulter automation systems. These tips are produced according to rigorous specifications and under stringent process controls, with automated assembly to ensure consistent performance and quality. They are certified to be free of RNase, DNase, and pyrogens. Available options include nonsterile, sterile, non-filtered, filtered, wide-bore, fine-point, and extended length versions. Compatible with Beckman Coulter Biomek FX, Biomek NX, Biomek 3000, Multimek AP-96, i-Series, SAP96 Head, Span-8, P20, MP20, P200L, and MP200. Filtered tips help minimize the risk of cross-contamination from aerosols. Like all Alkali Scientific™ tips, the P20s are made from 100% polypropylene, ensuring they are straight, leak-proof, and free from DNA, RNase/DNase, PCR inhibitors, pyrogens/endotoxins, and trace metals.
Catalog No. 50-302-6947 Supplier Alkali Scientific Supplier No. FXF10CS
Crafted from high-quality polypropylene, Alkali Scientific™ provides a wide range of tips for Beckman Coulter automation systems. These tips are produced according to rigorous specifications and under stringent process controls, with automated assembly to ensure consistent performance and quality. They are certified to be free of RNase, DNase, and pyrogens. Available options include nonsterile, sterile, non-filtered, filtered, wide-bore, fine-point, and extended length versions. Compatible with Beckman Coulter Biomek FX, Biomek NX, Biomek 3000, Multimek AP-96, i-Series, SAP96 Head, Span-8, P20, MP20, P200L, and MP200. Filtered tips help minimize the risk of cross-contamination from aerosols. Like all Alkali Scientific™ tips, the P20s are made from 100% polypropylene, ensuring they are straight, leak-proof, and free from DNA, RNase/DNase, PCR inhibitors, pyrogens/endotoxins, and trace metals.