Learn More
Alkali Scientific Alkali Scientific™ 10uL Beckman™ Biomek FX/NX, Multimek and i Series Compatible Robotic Automation Black Conductive Tips, Filtered 96-Well Head/Tip Format, Low Retention, Sterile, 10 Racks of 96, 960/Pack
SDP
Catalog No. 503027366
Encompass_Preferred
Crafted from high-quality polypropylene, Alkali Scientific™ offers a complete selection of tips for Beckman Coulter automation systems. These black conductive tips are produced according to strict specifications and under tight process controls, with automated assembly to ensure consistent quality and performance. Certified to be free of RNase, DNase, and pyrogens, they are available in various types, including nonsterile, sterile, non-filtered, filtered, wide-bore, fine-point, and extended length. These tips are compatible with Beckman Coulter Biomek FX, Biomek NX, Biomek 3000, Multimek AP-96, i-Series, SAP96 Head, Span-8, P20, MP20, P200L, and MP200. Filtered tips minimize the risk of sample cross-contamination from aerosols. Like all Alkali Scientific™ tips, the P20s are made from 100% polypropylene, ensuring they are straight, leak-proof, and free from DNA, DNase/RNase, PCR inhibitors, pyrogens/endotoxins, and trace metals.
Catalog No. 50-302-7366 Supplier Alkali Scientific Supplier No. FXBF10
Crafted from high-quality polypropylene, Alkali Scientific™ offers a complete selection of tips for Beckman Coulter automation systems. These black conductive tips are produced according to strict specifications and under tight process controls, with automated assembly to ensure consistent quality and performance. Certified to be free of RNase, DNase, and pyrogens, they are available in various types, including nonsterile, sterile, non-filtered, filtered, wide-bore, fine-point, and extended length. These tips are compatible with Beckman Coulter Biomek FX, Biomek NX, Biomek 3000, Multimek AP-96, i-Series, SAP96 Head, Span-8, P20, MP20, P200L, and MP200. Filtered tips minimize the risk of sample cross-contamination from aerosols. Like all Alkali Scientific™ tips, the P20s are made from 100% polypropylene, ensuring they are straight, leak-proof, and free from DNA, DNase/RNase, PCR inhibitors, pyrogens/endotoxins, and trace metals.