Learn More
Optimize Technologies OPTI-LYNX™ Trap Cartridges, 10µL Bed, C18AQ/40µm, 5pk
SDP
Catalog No. 502006291
Encompass_Preferred
OPTI-LYNX™ Trap Cartridges combine the convenience of our quick-connect holder with a versatile selection of packed-bed cartridges to offer chromatographers numerous options for on-line or off-line sample clean-up and pre-concentration. Each cartridge shares the same external dimensions, allowing you to use different diameters and porosities with the same holder hardware. OPTI-LYNX™ Traps are available in packed-bed volumes of 4 µL to 100 µL with a wide range of standard and custom packings. Whether you want to separate a peptide digest from its matrix for further analyses, or prepare a dilute small molecule sample for LC injection without any sample loss - OPTI-LYNX™ Traps are the ideal tool to optimize your technique. These columns can be loaded and regenerated repeatedly for maximum value.
Catalog No. 50-200-6291 Supplier Optimize Technologies Supplier No. 1103787TD
OPTI-LYNX™ Trap Cartridges combine the convenience of our quick-connect holder with a versatile selection of packed-bed cartridges to offer chromatographers numerous options for on-line or off-line sample clean-up and pre-concentration. Each cartridge shares the same external dimensions, allowing you to use different diameters and porosities with the same holder hardware. OPTI-LYNX™ Traps are available in packed-bed volumes of 4 µL to 100 µL with a wide range of standard and custom packings. Whether you want to separate a peptide digest from its matrix for further analyses, or prepare a dilute small molecule sample for LC injection without any sample loss - OPTI-LYNX™ Traps are the ideal tool to optimize your technique. These columns can be loaded and regenerated repeatedly for maximum value.