Learn More
SAI Infusion Technologies 10ul Untreated Capillary Blood Collection Tube
SDP
Catalog No. NC3353729
Encompass
10ul Untreated (Neutral) Microvolume Capillary Blood Collection Tube. Collect 10ul of blood via capillary action, then use the unique integrated plunger to dispense the sample quickly and cleanly. 200/box. Product is for laboratory animal research only.
Catalog No. NC3353729 Supplier SAI Infusion Technologies Supplier No. POCTN10
10ul Untreated (Neutral) Microvolume Capillary Blood Collection Tube. Collect 10ul of blood via capillary action, then use the unique integrated plunger to dispense the sample quickly and cleanly. 200/box. Product is for laboratory animal research only.