Learn More
Trajan Scientific VICI Valco Sample Loop 10-L Cheminert (VAL.CSL10)
Catalog No. 50635005
Encompass
VICI Valco Stainless Steel Loop With 1/16" Tube Ends Includes Two Stainless Steel Nuts and ferrµles. VICI Valco Part Number: CSL10 (NEW LPP PN: VAL.CSL10). Thermo Part Number: CH-953583. Made in the USA.
Catalog No. 50-635-005 Supplier Trajan Scientific Supplier No. PAL.CSL10
Add to Cart
Add to Cart
VICI Valco Stainless Steel Loop With 1/16" Tube Ends Includes Two Stainless Steel Nuts and ferrµles. VICI Valco Part Number: CSL10 (NEW LPP PN: VAL.CSL10). Thermo Part Number: CH-953583. Made in the USA.