Learn More
CELLTREAT Scientific Products 10L Extended Length Pipette Tips, Bulk Packed, Non-sterile
SDP
Catalog No. 501656725
Encompass_Preferred
Pipette Tips deliver high performance with exceptional value. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting. Extended length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels. Molded volume reference markings offer a quick double check opportunity. Bulk, non-sterile option available in unique self-standing, resealable bags. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed bulk non-sterile, 1000/re-sealable bag, 5000/case.
Catalog No. 50-165-6725 Supplier CELLTREAT Scientific Products Supplier No. 229043
Pipette Tips deliver high performance with exceptional value. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting. Extended length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels. Molded volume reference markings offer a quick double check opportunity. Bulk, non-sterile option available in unique self-standing, resealable bags. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed bulk non-sterile, 1000/re-sealable bag, 5000/case.