Learn More
Bellco Glass, Inc. 10 Extended Length Pipette
SDP
Catalog No. 502330684
Encompass_Preferred
CELLTREAT Bulk nonsterile Pipette Tips available in unique selfstanding resealable bags. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth surfaces to ensure accurate pipetting.Specifications Multiple sizes are available including extended length tips 10 and 100 to allow better access to the bottom of test tubes reagent bottles flasks and other vessels Molded volume reference markings offer a quick double check opportunity Case Quantity 5000
Catalog No. 50-233-0684 Supplier Bellco Glass, Inc. Supplier No. 229043000
CELLTREAT Bulk nonsterile Pipette Tips available in unique selfstanding resealable bags. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth surfaces to ensure accurate pipetting.Specifications Multiple sizes are available including extended length tips 10 and 100 to allow better access to the bottom of test tubes reagent bottles flasks and other vessels Molded volume reference markings offer a quick double check opportunity Case Quantity 5000