Learn More
Quality Biological Inc 10 µl Inoculating Loop, Individual Pack, Blue, 221mm, Sterile
SDP
Catalog No. 50147468
Encompass_Preferred
10 µl Inoculating Loop, Individual Pack, Blue, Sterile
Packaged 1/Wrap, 600/CS
• Flexible for easy access to small volume tubes and dishes
• Color coded for easy identification
• Gentle on delicate agar or other surfaces
Catalog No. 50-147-468 Supplier Quality Biological Inc Supplier No. P886229612
10 µl Inoculating Loop, Individual Pack, Blue, Sterile
Packaged 1/Wrap, 600/CS
• Flexible for easy access to small volume tubes and dishes
• Color coded for easy identification
• Gentle on delicate agar or other surfaces