Learn More
Trajan Scientific L-MARK Brand 10l Syringe, Gold, FN, GT, Pt. 3, 22s, Coated NDL
Catalog No. 50635268
Encompass_Preferred
L-MARK Brand Syringe, 10ul, Fixed Needle, FN, Chemically Inert Coated Needle, GT, PTFE, 22s Gauge, Pt. Style 3 Made in Germany.
Catalog No. 50-635-268 Supplier Trajan Scientific Supplier No. LMK.2606416C
L-MARK Brand Syringe, 10ul, Fixed Needle, FN, Chemically Inert Coated Needle, GT, PTFE, 22s Gauge, Pt. Style 3 Made in Germany.