Learn More
Trajan Scientific L-MARK Brand 10l Syringe, FN, GT, Pt. 3, 22s
Catalog No. 50635263
Encompass_Preferred
L-MARK Brand 10l, Syringe, FN, GT, 22gs, NDL Length 51mm, Pt. Style 3, Plunger Tip PTFE, Single Made in Germany.
Catalog No. 50-635-263 Supplier Trajan Scientific Supplier No. LMK.2606416
L-MARK Brand 10l, Syringe, FN, GT, 22gs, NDL Length 51mm, Pt. Style 3, Plunger Tip PTFE, Single Made in Germany.