Learn More
Waters Corp Waters Sample Loop, 10 μL, Stainless Steel
Catalog No. NC9530355 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 10 μL sample loop is designed to fit into the 5000psi nanoACQUITY system and is also utilized on the Open Architecture UPLC System.
  • Capacity of 10 μL.
  • Constructed from stainless steel.
  • Compatible with 5000psi nanoACQUITY systems.
  • Compatible with Open Architecture UPLC systems.
  • Used for UPLC system applications.
Catalog No. NC9530355 Supplier Waters Corp Supplier No. 430001326
The 10 μL sample loop is designed to fit into the 5000psi nanoACQUITY system and is also utilized on the Open Architecture UPLC System.
  • Capacity of 10 μL.
  • Constructed from stainless steel.
  • Compatible with 5000psi nanoACQUITY systems.
  • Compatible with Open Architecture UPLC systems.
  • Used for UPLC system applications.