Learn More
Bellco Glass, Inc. 10uL low retention filter
SDP
Supplier:  Bellco Glass, Inc. 229015000
Encompass_Preferred
Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing process results in superior low retentionPipette Tips are universal fit and compatible with major pipettesPipette Tips have smooth, hydrophobic surfaces to ensure accurate pipettingFilters reduce cross contamination and prevent damaging aerosols from penetrating into the pipette, and are positioned to allow extra headspace when pipetting maximum volumeMolded volume reference markings offer a quick double check opportunityPipette Tip RacksHinged Lids are designed for easy one-handed opening, making it easier to access repeatedlyLids have a stacking feature for added stability and a secure locking tabInterior support ribs prevent rack from warping Pack Description96 Tips/RackCase Quantity960SterileYesRNase/DNase FreeYesAutoclavableNoLow RetentionYesFilterYesRackedYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume10µLGraduatedYes
Catalog No. 50-232-9450