Learn More
AMPLYUS LLC miniPCR bio 0.5-10 µl micropipette tips – 2 racks of 96
SDP
Catalog No. NC1974917
Encompass_Preferred
These clear micropipette tips are the perfect companion for any standard 0.5-10 µl micropipette. Universal fit, 2 racks of 96 tips each. Tips are autoclavable and certified to be free of any detectable RNase, DNase, DNA, or other contaminants.
Catalog No. NC1974917 Supplier AMPLYUS LLC Supplier No. CM100105
These clear micropipette tips are the perfect companion for any standard 0.5-10 µl micropipette. Universal fit, 2 racks of 96 tips each. Tips are autoclavable and certified to be free of any detectable RNase, DNase, DNA, or other contaminants.