Learn More
Genesee Scientific Corporation Olympus 10 µl Reach Barrier Tips, Low-Retention, RNase/DNase Free, Sterile, 960 Tips/Unit
SDP
Catalog No. NC1962411
Encompass
The Olympus Plastics 10 µl Reach Barrier Tips are precision-engineered, sterile pipette tips designed for critical molecular biology applications. They feature a positive sealing barrier to prevent contamination and are RNase, DNase, DNA, and pyrogen-free certified. Their low-retention design enhances accuracy and minimizes sample loss, making them ideal for sensitive assays. Manufactured from virgin polypropylene, these tips offer universal compatibility with various pipettors, ensuring reliable performance in the lab.
Catalog No. NC1962411 Supplier Genesee Scientific Corporation Supplier No. 26401
The Olympus Plastics 10 µl Reach Barrier Tips are precision-engineered, sterile pipette tips designed for critical molecular biology applications. They feature a positive sealing barrier to prevent contamination and are RNase, DNase, DNA, and pyrogen-free certified. Their low-retention design enhances accuracy and minimizes sample loss, making them ideal for sensitive assays. Manufactured from virgin polypropylene, these tips offer universal compatibility with various pipettors, ensuring reliable performance in the lab.