Learn More
Chemglass Life Sciences Chemglass Pipette Tips, 10 μL, Clear, Bulk, Non-sterile - 1,000/Pack, 10,000/Case
SDP
Catalog No. 502733211
Encompass_Preferred
Chemglass 10 μL clear pipette tips are bulk-packed, non-sterile polypropylene tips designed for routine low-volume liquid handling. Ultra-clear material provides visual accuracy while a low-retention fine point minimizes sample adhesion and improves recovery.
  • Nominal 10 μL volume for small-volume dispensing.
  • Ultra-clear polypropylene resists organic solvents and improves visibility.
  • Low-retention fine-point design reduces sample adhesion.
  • Non-sterile, bulk-packed for high-throughput workflows (1,000/pack; 10,000/case).
  • Universal fit compatible with most single- and multichannel pipettors.
  • Certified free of RNase, DNase, human DNA, and pyrogens where applicable.
Catalog No. 50-273-3211 Supplier Chemglass Life Sciences Supplier No. CLS3499E018
Chemglass 10 μL clear pipette tips are bulk-packed, non-sterile polypropylene tips designed for routine low-volume liquid handling. Ultra-clear material provides visual accuracy while a low-retention fine point minimizes sample adhesion and improves recovery.
  • Nominal 10 μL volume for small-volume dispensing.
  • Ultra-clear polypropylene resists organic solvents and improves visibility.
  • Low-retention fine-point design reduces sample adhesion.
  • Non-sterile, bulk-packed for high-throughput workflows (1,000/pack; 10,000/case).
  • Universal fit compatible with most single- and multichannel pipettors.
  • Certified free of RNase, DNase, human DNA, and pyrogens where applicable.