Learn More
Andwin Scientific 10UL PIPETTE TIPS
SDP
Catalog No. NC9961024
Encompass_Preferred
The 0.110 L pipette tips are compatible with most micropipettors. The tips are sterile and are free of endotoxins. This product is autoclavable up to 122C (252F). The tips measure 3.1 cm (1.235") long.
Catalog No. NC9961024 Supplier Andwin Scientific Supplier No. 53509132
The 0.110 L pipette tips are compatible with most micropipettors. The tips are sterile and are free of endotoxins. This product is autoclavable up to 122C (252F). The tips measure 3.1 cm (1.235") long.