Learn More
CELLTREAT Scientific Products 10L Pipette Tips, Bulk Packed, Non-sterile
SDP
Catalog No. 501656724
Encompass_Preferred
Pipette Tips deliver high performance with exceptional value. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting. Molded volume reference markings offer a quick double check opportunity. Bulk, non-sterile option available in unique self-standing, resealable bags. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed bulk non-sterile, 1000/re-sealable bag, 5000/case.
Catalog No. 50-165-6724 Supplier CELLTREAT Scientific Products Supplier No. 229042
Pipette Tips deliver high performance with exceptional value. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth, hydrophobic surfaces to ensure accurate pipetting. Molded volume reference markings offer a quick double check opportunity. Bulk, non-sterile option available in unique self-standing, resealable bags. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packed bulk non-sterile, 1000/re-sealable bag, 5000/case.