Learn More
Trajan Scientific Hamilton 10l Syringe, FN, 23s, Fitted, Pt. AS
Catalog No. 50711784
Encompass_Preferred
Hamilton 10¿l, Hamilton Syringe FN, Fitted, 23s Gauge, NDL Length 43.4mm, Pt. Style AS, Plunger Tip Fitted, 6 Pack
Catalog No. 50-711-784 Supplier Trajan Scientific Supplier No. HAM.80390
Hamilton 10¿l, Hamilton Syringe FN, Fitted, 23s Gauge, NDL Length 43.4mm, Pt. Style AS, Plunger Tip Fitted, 6 Pack