Learn More
Thermo Scientific GC Syringes for TriPlus RSH Autosampler | | 1 EA
Catalog No. NC2160487 Shop All Thermo Scientific Products
Encompass
Achieve accurate precise and reproducible injections easily using Thermo Scientific GC Syringes for Thermo Scientific TriPlus RSH Autosampler. The robust field-proven designs are optimized for septum piercing. They are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass and cover a range of volumes. Syringe volumes from 0.5μL to 5mL. Gas-tight options available.
Catalog No. NC2160487 Supplier Thermo Scientific Supplier No. 365D0291
Achieve accurate precise and reproducible injections easily using Thermo Scientific GC Syringes for Thermo Scientific TriPlus RSH Autosampler. The robust field-proven designs are optimized for septum piercing. They are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass and cover a range of volumes. Syringe volumes from 0.5μL to 5mL. Gas-tight options available.