Learn More
Chemglass Life Sciences Syringe, Micro, 10uL Capacity, Removable Needle, 26s Gauge
SDP
Catalog No. 50872574
Encompass_Preferred
Syringe, Micro, 10uL Capacity, Removable Needle, 26s Gauge. Basic micro syringe used for the precise liquid delivery in volumes of 10 to 100uL. The accuracy of these syringes is ± 1% of the total volume with the repeatability of dispensed volume being ± 1%. This precision and accuracy is a direct result of the close tolerances of the stainless steel plunger rod, custom shrinking and polishing of the glass bore. Each syringe plunger is individually hand fitted, assembled into the glass bore and tested to be liquid tight to 80 PSI. The plungers and barrels are not interchangeable.
Catalog No. 50-872-574 Supplier Chemglass Life Sciences Supplier No. CG307204
Syringe, Micro, 10uL Capacity, Removable Needle, 26s Gauge. Basic micro syringe used for the precise liquid delivery in volumes of 10 to 100uL. The accuracy of these syringes is ± 1% of the total volume with the repeatability of dispensed volume being ± 1%. This precision and accuracy is a direct result of the close tolerances of the stainless steel plunger rod, custom shrinking and polishing of the glass bore. Each syringe plunger is individually hand fitted, assembled into the glass bore and tested to be liquid tight to 80 PSI. The plungers and barrels are not interchangeable.