Learn More
Genesee Scientific Corporation Genesee Scientific 10µL XL Barrier Tips, RNase, DNase, DNA and Pyrogen-Free, Universal Compatibility, 10 µL
SDP
Catalog No. NC2400186
Encompass
The Genesee Scientific Olympus 10µL XL Premium Barrier Tips are designed for high-precision liquid handling in sensitive scientific environments. Crafted from virgin polypropylene, these sterile tips minimize sample retention and adhesion of DNA and proteins, ensuring reliable results. Featuring a low-binding, hydrophobic surface and universal compatibility with most research-grade pipettors, they stand out for their no silicone additives and broad chemical resistance, making them ideal for diverse laboratory applications.
Catalog No. NC2400186 Supplier Genesee Scientific Corporation Supplier No. 23401
The Genesee Scientific Olympus 10µL XL Premium Barrier Tips are designed for high-precision liquid handling in sensitive scientific environments. Crafted from virgin polypropylene, these sterile tips minimize sample retention and adhesion of DNA and proteins, ensuring reliable results. Featuring a low-binding, hydrophobic surface and universal compatibility with most research-grade pipettors, they stand out for their no silicone additives and broad chemical resistance, making them ideal for diverse laboratory applications.