Learn More
Andwin Scientific 10UL YL INC LOOP 1000/CS
SDP
Catalog No. NC9504018
Encompass_Preferred
Made with high-impact polystrene, the inoculating loops feature uniform and smooth streaking without damaging the gel surface. The product is sterile and is packaged in a secure, tamperproof, zip-seal resealable bag. The loops are disposable and do not require flaming, which removes the chance of infection from the formation of aerosol. This product comes in a case of 1000 yellow loops.
Catalog No. NC9504018 Supplier Andwin Scientific Supplier No. 12000810
Made with high-impact polystrene, the inoculating loops feature uniform and smooth streaking without damaging the gel surface. The product is sterile and is packaged in a secure, tamperproof, zip-seal resealable bag. The loops are disposable and do not require flaming, which removes the chance of infection from the formation of aerosol. This product comes in a case of 1000 yellow loops.