Learn More
Bellco Glass, Inc. 10 /10 Extended Length
SDP
Catalog No. 502330794
Encompass_Preferred
Hinged Lids are designed for easy onehanded opening making it easier to access repeatedly. Reduce plastic waste by refilling empty racks available without a wafer to allow refill from pipette tip reload systems.Specifications Lids have a stacking feature for added stability and a secure locking tab Interior support ribs prevent rack from warping Case Quantity 20
Catalog No. 50-233-0794 Supplier Bellco Glass, Inc. Supplier No. 229060000
Hinged Lids are designed for easy onehanded opening making it easier to access repeatedly. Reduce plastic waste by refilling empty racks available without a wafer to allow refill from pipette tip reload systems.Specifications Lids have a stacking feature for added stability and a secure locking tab Interior support ribs prevent rack from warping Case Quantity 20