Learn More
Bellco Glass, Inc. 10 /10 Extended Length
Supplier: Bellco Glass, Inc. 229065000
Empty racks are available with a wafer to allow refill from bulk packed pipette tips. Hinged Lids are designed for easy onehanded opening making it easier to access repeatedly.Specifications Lids have a stacking feature for added stability and a secure locking tab Interior support ribs prevent rack from warping Case Quantity 20