Learn More
Alkali Scientific 100 um Cell Strainer, PP, Gamma Sterilized, 50/pk
SDP
Catalog No. 502335801
Encompass_Preferred
ASI™ Cell Strainers make obtaining a more uniform single cell easy. Individually packaged and gamma sterilized. ASI cell strainers are the perfect choice for your research. • Individually Wrapped Pre-sterilized • Reduced Chance of Contamination • Nylon Mesh Filtration • Fits Standard 50mL Conical Tubes • Diameter bottom: 27.05mm, Height:14.50 mm • SAL 10^-6 sterilization • QTY: 50 Units/Case
Catalog No. 50-233-5801 Supplier Alkali Scientific Supplier No. TS100
ASI™ Cell Strainers make obtaining a more uniform single cell easy. Individually packaged and gamma sterilized. ASI cell strainers are the perfect choice for your research. • Individually Wrapped Pre-sterilized • Reduced Chance of Contamination • Nylon Mesh Filtration • Fits Standard 50mL Conical Tubes • Diameter bottom: 27.05mm, Height:14.50 mm • SAL 10^-6 sterilization • QTY: 50 Units/Case