Learn More
Trilink Bio Technologies Inc N1-Methyl-pseudouridine 5'-triphosphate
SDP
Catalog No. NC1443155
Encompass
N1-Methylpseudouridine 5'-Triphosphate is a modified nucleoside triphosphate used in RNA research and synthesis. It contains a methyl group on the N1 position of the pseudouridine nucleobase. This analog is cell permeable and can be incorporated into RNA by polymerases. It is supplied as a lithium salt at ≥95% purity by TriLink BioTechnologies. The product has a molecular weight of 491.15 g/mol and is soluble up to 100 mM in water or buffer. It can be used to study RNA structure and function.
Catalog No. NC1443155 Supplier Trilink Bio Technologies Inc Supplier No. N108110
N1-Methylpseudouridine 5'-Triphosphate is a modified nucleoside triphosphate used in RNA research and synthesis. It contains a methyl group on the N1 position of the pseudouridine nucleobase. This analog is cell permeable and can be incorporated into RNA by polymerases. It is supplied as a lithium salt at ≥95% purity by TriLink BioTechnologies. The product has a molecular weight of 491.15 g/mol and is soluble up to 100 mM in water or buffer. It can be used to study RNA structure and function.