Learn More
Diagnostic Biosystems 10X ANTIGEN RETRIEVAL 10X Antigen Retrieval Buffer High pH 10.0
Catalog No. 502407621
Encompass_Preferred
10X Antigen Retrieval Buffer High pH 10.0 is designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin embedded tissue sections. The use of this buffer in combination with heat (often by microwave, water bath, or pressure cooker) has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue. This buffer is supplied as a 10X stock solution.
Catalog No. 50-240-7621 Supplier Diagnostic Biosystems Supplier No. K044
10X Antigen Retrieval Buffer High pH 10.0 is designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin embedded tissue sections. The use of this buffer in combination with heat (often by microwave, water bath, or pressure cooker) has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue. This buffer is supplied as a 10X stock solution.