Learn More
Apexbio Technology LLC 10X Binding Buffer 10ml
SDP
Catalog No. 503107160
Encompass_Preferred
This reagent is an auxiliary reagent for Annexin V apoptosis detection and can be used in combination with fluorescein-labeled Annexin V to detect cell apoptosis
Catalog No. 50-310-7160 Supplier Apexbio Technology LLC Supplier No. K228410
This reagent is an auxiliary reagent for Annexin V apoptosis detection and can be used in combination with fluorescein-labeled Annexin V to detect cell apoptosis