Learn More
Abcam 10 x Blocking Buffer, 50ML
Catalog No. NC0481068
Encompass_Preferred
10X blocking buffer (pH 7.5) for serum-free blocking of non-specific antibody binding. - Cited in over 30 publications - Suitable for ELISA, in-cell ELISA, ICC/IF, western blot (WB) - 100% casein blocking as preservative
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0481068 Supplier Abcam Supplier No. AB12658750ML
10X blocking buffer (pH 7.5) for serum-free blocking of non-specific antibody binding. - Cited in over 30 publications - Suitable for ELISA, in-cell ELISA, ICC/IF, western blot (WB) - 100% casein blocking as preservative
The product is subject to the following: Abcam Restricted Use Statement