Learn More
Neuromics, Inc 10x Buffer BXT, Strep-Tacting XT Elution Buffer, 25 ml contains a Biotin concentration for competitve displacement of purified recombinant proteins.
Catalog No. NC2071873
Encompass
Buffer BXT is a Strep-TactinXT Elution Buffer with Biotin. It contains a Biotin concentration for competitive displacement of purified recombinant Strep-tagII or Twin-Strep-tag fusion proteins from immobilized Strep-TactinXT.Buffer BXT is also recommended for efficient elution in combination with Strep-TactinXT and MagStrep "type3" XT beads (2-4090-002).
Catalog No. NC2071873 Supplier Neuromics, Inc Supplier No. 21042025
Buffer BXT is a Strep-TactinXT Elution Buffer with Biotin. It contains a Biotin concentration for competitive displacement of purified recombinant Strep-tagII or Twin-Strep-tag fusion proteins from immobilized Strep-TactinXT.Buffer BXT is also recommended for efficient elution in combination with Strep-TactinXT and MagStrep "type3" XT beads (2-4090-002).