Learn More
IBA LifeSciences 10x Buffer BXT - Strep-TactinXT Elution Buffer with Biotin; 25 ml
Catalog No. NC1445684
Encompass_Preferred
10x Buffer BXT is a 10x concentrated Strep-TactinXT elution buffer containing biotin. Biotin is a water-soluble B-complex vitamin, also known as vitamin B7 or vitamin H. It binds to Strep-TactinXT with very high affinity but reversibly and thereby allows the efficient elution of Strep-tagII or Twin-Strep-tag fusion proteins from the resin. Please note that 10x Buffer BXT is not applicable for elution of Strep-tagII or Twin-Strep-tag fusion proteins from Strep-Tactin, since biotin irreversibly binds to these resins, and thus Strep-Tactin cannot be regenerated.
Catalog No. NC1445684 Supplier IBA LifeSciences Supplier No. 21042025
10x Buffer BXT is a 10x concentrated Strep-TactinXT elution buffer containing biotin. Biotin is a water-soluble B-complex vitamin, also known as vitamin B7 or vitamin H. It binds to Strep-TactinXT with very high affinity but reversibly and thereby allows the efficient elution of Strep-tagII or Twin-Strep-tag fusion proteins from the resin. Please note that 10x Buffer BXT is not applicable for elution of Strep-tagII or Twin-Strep-tag fusion proteins from Strep-Tactin, since biotin irreversibly binds to these resins, and thus Strep-Tactin cannot be regenerated.