Learn More
Sony Biotechnology 10x ClearSort Sheath Fluid, Sterile, 4 x 1L
Catalog No. NC0439131
Encompass_Preferred
The 10x ClearSort Sheath Fluid is a sterile, ready-to-use sheath fluid for flow cytometry applications. It is optimized for use with 10x Genomics' single cell platforms including the Chromium Controller, Chromium Single Cell Controller, and Chromium Single Cell A Chip Kit. The sheath fluid contains a proprietary mix of polymers and salts formulated to gently deliver cells into droplets while maintaining cell viability. It is manufactured under cGMP conditions and 0.2 micron sterile-filtered. The fluid is available in 1L bottles, sold in cases of 4. It has a 12 month shelf life when stored properly. The fluid is compatible with most cell types including PBMCs, immune cells, and cancer cells.
Catalog No. NC0439131 Supplier Sony Biotechnology Supplier No. AE700713
The 10x ClearSort Sheath Fluid is a sterile, ready-to-use sheath fluid for flow cytometry applications. It is optimized for use with 10x Genomics' single cell platforms including the Chromium Controller, Chromium Single Cell Controller, and Chromium Single Cell A Chip Kit. The sheath fluid contains a proprietary mix of polymers and salts formulated to gently deliver cells into droplets while maintaining cell viability. It is manufactured under cGMP conditions and 0.2 micron sterile-filtered. The fluid is available in 1L bottles, sold in cases of 4. It has a 12 month shelf life when stored properly. The fluid is compatible with most cell types including PBMCs, immune cells, and cancer cells.