Learn More
Leinco Technologies Inc 10X ClearFlow™ Sheath Fluid | 20L
SDP
Catalog No. NC1437741
Encompass_Preferred
10X ClearFlow™ Sheath Fluid is specifically formulated for transporting particles through a flow cytometer. ClearFlow™ is specially manufactured in our class 10,000 clean room environment to assure low particulate counts and low fluorescence backgrounds.
Catalog No. NC1437741 Supplier Leinco Technologies Inc Supplier No. S62220L
10X ClearFlow™ Sheath Fluid is specifically formulated for transporting particles through a flow cytometer. ClearFlow™ is specially manufactured in our class 10,000 clean room environment to assure low particulate counts and low fluorescence backgrounds.