Learn More
Sigma Aldrich Fine Chemicals Biosciences 10X CRISPRi feature Barcod
Catalog No. 502905715
Encompass
This kit includes pooled virus representing each of the 2 possible capture sequences and 2 position combinations (capture sequences 1 or 2 in the stem-loop or 3 position). Present within each pool are 2 CRISPRi sgRNAs 1 targeting RAB1A and 1 non-targeting control sgRNA. Each pool is provided ready to use at 1x106 viral titer sufficient enough to cover 2x106 cells at an MOI of 0.5. The purpose of the optimization kit is to set up a pilot screen to determine the best capture sequence and position configuration before a custom pool is ordered.We have an exclusive partnership with 10x Genomics our compatible CRISPRi pools are a perfect complementary product for target gene validation following a whole genome screen.The 10X CRISPRi feature Barcode Optimization kit is ideally suited for setting up the screen assay and if desired capture sequence optimization before using the Custom 10x Compatible CRISPRi library pools. for more information about single cell screening with CRISPR and 10x
Catalog No. 50-290-5715 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CRISPRI10X1KT
This kit includes pooled virus representing each of the 2 possible capture sequences and 2 position combinations (capture sequences 1 or 2 in the stem-loop or 3 position). Present within each pool are 2 CRISPRi sgRNAs 1 targeting RAB1A and 1 non-targeting control sgRNA. Each pool is provided ready to use at 1x106 viral titer sufficient enough to cover 2x106 cells at an MOI of 0.5. The purpose of the optimization kit is to set up a pilot screen to determine the best capture sequence and position configuration before a custom pool is ordered.We have an exclusive partnership with 10x Genomics our compatible CRISPRi pools are a perfect complementary product for target gene validation following a whole genome screen.The 10X CRISPRi feature Barcode Optimization kit is ideally suited for setting up the screen assay and if desired capture sequence optimization before using the Custom 10x Compatible CRISPRi library pools. for more information about single cell screening with CRISPR and 10x