Learn More
Nanotemper Technologies Inc 10X DIANTHUS MICROWELL PLATE
Catalog No. NC1846255
Encompass
  • Microwell plate type for biophysical assays.
  • Features 384 wells for high-throughput screening.
  • Plates are barcoded for easy sample tracking.
  • Compatible with Dianthus system for TRIC measurements.
  • Constructed from black/black polystyrene for light protection.
  • Clear bottom for optical detection from below.
  • Working volume range of 18-25 µL to suit various assays.
  • Optimal working volume of 20 µL for enhanced performance.
  • Coated with a special polymer to minimize protein adsorption.
  • Packs include a rack of 10 plates for convenient use.
Catalog No. NC1846255 Supplier Nanotemper Technologies Inc Supplier No. DIP001B
  • Microwell plate type for biophysical assays.
  • Features 384 wells for high-throughput screening.
  • Plates are barcoded for easy sample tracking.
  • Compatible with Dianthus system for TRIC measurements.
  • Constructed from black/black polystyrene for light protection.
  • Clear bottom for optical detection from below.
  • Working volume range of 18-25 µL to suit various assays.
  • Optimal working volume of 20 µL for enhanced performance.
  • Coated with a special polymer to minimize protein adsorption.
  • Packs include a rack of 10 plates for convenient use.