Learn More
Teknova 10X DPBS Concentrate without Calcium and Magnesium Salts. pH 7.4. 1Liter Bottle. Non Sterile. [H0] 2 pack.
SDP
Catalog No. 502387606
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
Catalog No. 50-238-7606 Supplier Teknova Supplier No. P02952PK
Add to Cart
Add to Cart
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.