Learn More
Quality Biological Inc DPBS 10X without Calcium and Magnesium, 4 Liters
SDP
Catalog No. 50146768
Encompass_Preferred
DPBS is a balanced salt solution ¿ Used for the handling and culturing of mammalian cells. ¿ DPBS without Ca and Mg can be used to wash and rinse suspended cells. ¿ Without Phenol Red ¿ Serves as an irrigating, transporting or diluting fluid for mammalian cells, while maintaining cell tonicity and viability for a limited period of time. ¿ Diluent of fresh red blood cells to study the trapping of red blood cell aggregate ¿ Wash the wells of microtiter plates. ¿ Storage Condition: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Endotoxin and Mycoplasma Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 12161700 ¿ ¿ Max Shelf Life: 24 months
Catalog No. 50-146-768 Supplier Quality Biological Inc Supplier No. 119068491
Add to Cart
Add to Cart
DPBS is a balanced salt solution ¿ Used for the handling and culturing of mammalian cells. ¿ DPBS without Ca and Mg can be used to wash and rinse suspended cells. ¿ Without Phenol Red ¿ Serves as an irrigating, transporting or diluting fluid for mammalian cells, while maintaining cell tonicity and viability for a limited period of time. ¿ Diluent of fresh red blood cells to study the trapping of red blood cell aggregate ¿ Wash the wells of microtiter plates. ¿ Storage Condition: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Endotoxin and Mycoplasma Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 12161700 ¿ ¿ Max Shelf Life: 24 months