Learn More
Seracare Life Sciences Inc 10X DULBECCO S PBS (D-PBS)
Catalog No. 5067481
Encompass_Preferred
10X Phosphate-buffered Saline (PBS) is ideal for use in sample preparation and as a wash buffer in general immunoassay applications as well as tissue and cell culture procedures. This modified Dulbecco's buffer is not formulated with magnesium or calcium salts. pH=7.4. Conductivity controlled.
This item is not returnable. View return policy
Catalog No. 50-674-81 Supplier Seracare Life Sciences Inc Supplier No. 54600031
10X Phosphate-buffered Saline (PBS) is ideal for use in sample preparation and as a wash buffer in general immunoassay applications as well as tissue and cell culture procedures. This modified Dulbecco's buffer is not formulated with magnesium or calcium salts. pH=7.4. Conductivity controlled.