Learn More
Seracare Life Sciences Inc 10X DULBECCO S PBS W/ TWEEN 20
Supplier:  Seracare Life Sciences Inc 54600034
Encompass_Preferred
10X Phosphate-buffered Saline (PBS) is ideal for use in sample preparation and as a wash buffer in general immunoassay applications as well as tissue and cell culture procedures. This modified Dulbecco's buffer is not formulated with magnesium or calcium salts. Tween 20, a non-ionic detergent, reduces non-specific binding and protein-protein interaction during the wash step in immunoassay procedures. pH=7.4. Conductivity controlled.
Catalog No. 50-674-83