Learn More
Fisherbrand™ 10x Eyepiece with Micrometer
GREENER_CHOICE
Catalog No. 03000147 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Microscope
Catalog No. For Use With (Equipment)
03-000-147 Microscope
1 options
Catalog No. 03-000-147
Add to Cart
Add to Cart
High eye point, wide field, flat field eyepieces with 1 mm interval micrometer scale.
High eye point, wide field, flat field eyepieces.
  • 1 mm interval micrometer scale
Specifications
Type Eyepiece
For Use With (Equipment) Microscope