Learn More
Teknova 10X HBS-P Buffer, pH 7.4 at 1X. 1000mL, Sterile
SDP
Catalog No. 502388184
Encompass
1.5M Sodium Chloride,0.1M HEPES,0.5% Tween-20
Catalog No. 50-238-8184 Supplier Teknova Supplier No. H8174
Add to Cart
Add to Cart
1.5M Sodium Chloride,0.1M HEPES,0.5% Tween-20